ContactUsLinkComponent
STAT5 A/B pY694/699 ELISA Kit
STAT5 A/B pY694/699 ELISA Kit
Catalog # ABCAAB176656-96
Supplier:  Abcam
CAS Number:  
undefined
STAT5 A/B pY694/699 ELISA Kit
Catalog # ABCAAB176656-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    STAT5 A/B pY694/699 ELISA Kit
  • Tests per kit:
    96 tests
Specifications
About this item
STAT5 A/B pY694/699 ELISA Kit is a single-wash 90-min Simplestep used to quantify STAT5 A/B pY694/699 with a sensitivity of 5 µg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.