Mouse IP-10 ELISA Kit (CXCL10)
Supplier: Abcam
Certificates
About this item
Mouse IP-10 ELISA kit (CXCL10) is a single-wash 90-min Simplestep used to quantify Mouse IP-10 (CXCL10) with a sensitivity of 21,75 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 384-well plate for higher throughput.
Specifications
- Storage temperature:+4 ℃