Human SPARC ELISA Kit
ABCAAB220654-96
: Abcam
:
- Description:Human SPARC ELISA kit
Human SPARC ELISA kit is a single-wash 90-min Simplestep used to quantify Human SPARC with a sensitivity of 115 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.