Mouse CXCL1 ELISA Kit (GRO alpha)
Catalog # ABCAAB216951-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse CXCL1 ELISA Kit (GRO alpha)
- Tests per kit:96 tests
Specifications
About this item
Mouse CXCL1 ELISA Kit (GRO alpha) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse CXCL1 (GRO alpha) in Mouse in Cell culture extracts, Heparin Plasma, Citrate plasma, Cell culture supernatant, Serum, EDTA Plasma samples.