Rat Plasminogen ELISA Kit
Catalog # ABCAAB157740-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat Plasminogen ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Rat Plasminogen ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat Plasminogen in Rat in Plasma, Serum samples.