ContactUsLinkComponent
Complex IV Human Enzyme Activity Dipstick Assay Kit
  ABCAAB109876-30
 :  Abcam
 :  
undefined
Complex IV Human Enzyme Activity Dipstick Assay Kit
  ABCAAB109876-30
 :  Abcam
 :  
  • Description:
    Complex IV human enzyme activity dipstick assay kit
  • Method:
    Sandwich
  • Protein:
    Complex IV
  • Species reactivity:
    Other;Human
  • Processing time:
    Multiple steps
  • Product type:
    Pre-coated
  • Tests per kit:
    30 tests
Complex IV Human Enzyme Activity Dipstick Assay Kit is an immunoassay kit for the detection of Complex IV Human Enzyme Activity in Cow, Human in Cell culture extracts, Tissue samples.