ContactUsLinkComponent
Human Complement C5 ELISA Kit
Human Complement C5 ELISA Kit
Catalog # ABCAAB125963-96
Supplier:  Abcam
CAS Number:  
Human Complement C5 ELISA Kit
Catalog # ABCAAB125963-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Human Complement C5 ELISA kit
  • Tests per kit:
    96 tests
Specifications
About this item
Human Complement C5 ELISA kit ab125963 is used to quantify Human Complement C5 with a sensitivity of 26,54 pg/ml. Sandwich ELISA kit.