Clostridioides difficile Toxin A Antibody Pair - BSA and Azide Free
Catalog # ABCAAB316224-10X96
Supplier: Abcam
New Product
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:Clostridium difficile Toxin A
- Description:Clostridioides difficile Toxin A antibody pair - BSA and azide free
- Detection range:78 - 5000 pg/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:10×96 tests
Specifications
About this item
Clostridioides difficile Toxin A antibody pair - BSA and Azide free.