Human Folate Binding Protein / FBP ELISA Kit
Catalog # ABCAAB213781-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Folate Binding Protein / FBP ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Human Folate Binding Protein / FBP ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Human Folate Binding Protein / FBP in Human in cell culture supernatant, EDTA Plasma, Heparin Plasma, Milk, Saliva, Serum, Urine samples.