Human JAM-A Antibody Pair - BSA and Azide Free (Junctional Adhesion Molecule)
Catalog # ABCAAB253466-10X96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:Junctional Adhesion Molecule 1/JAM-A
- Description:Human JAM-A antibody pair - BSA and azide free (Junctional Adhesion Molecule)
- Detection range:15,625 - 1000 pg/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:10×96 tests
Specifications
About this item
Human JAM-A antibody pair - BSA and Azide free (Junctional Adhesion Molecule).