ERK1 (phospho T202 + Y204) + ERK2 (phospho T185 + Y187) In-Cell ELISA Kit
ABCAAB126423-96
: Abcam
:
- Description:ERK1 (phospho T202 + Y204) + ERK2 (phospho T185 + Y187) In-Cell ELISA kit
ERK1 (phospho T202 + Y204) + ERK2 (phospho T185 + Y187) In-cell ELISA kit is cell-based (qualitative) ELISA for the measurement of ERK1 (phospho T202 + Y204) + ERK2 (phospho T185 + Y187) production in Mouse, Rat, Human in Adherent cells samples.