Human C9 ELISA Kit (Complement C9)
Catalog # ABCAAB287178-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human C9 ELISA kit (Complement C9)
- Tests per kit:96 tests
Specifications
About this item
Human C9 ELISA kit (Complement C9) is a single-wash 90-min Simplestep used to quantify Human C9 (Complement C9) with a sensitivity of 10,6 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair.