Mouse TIM 1 ELISA Kit (KIM-1)
Catalog # ABCAAB213477-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse TIM 1 ELISA kit (KIM-1)
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Mouse TIM 1 ELISA kit (KIM-1) is a single-wash 90-min Simplestep used to quantify Mouse TIM 1 (KIM-1) with a sensitivity of 1,6 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 384-well plate for higher throughput.