Human Apolipoprotein AI ELISA Kit (APOA1)
Catalog # ABCAAB108804-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Apolipoprotein AI ELISA kit (APOA1)
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Human Apolipoprotein AI ELISA kit (APOA1) is a Competitive ELISA kit for the measurement of Human Apolipoprotein AI (APOA1) in Human in plasma, serum samples.