Rat Fibrinogen ELISA Kit
ABCAAB108845-96
: Abcam
:
- Description:Rat Fibrinogen ELISA Kit
- Tests per kit:96 tests
Rat Fibrinogen ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat Fibrinogen in Rat in Cell Lysate, Cell culture supernatant, Tissue Lysate, Urine samples.