Phosphotyrosine Tec ELISA Kit
Catalog # ABCAAB279954-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Phosphotyrosine Tec ELISA kit
Specifications
About this item
Phosphotyrosine Tec ELISA kit is a Semi-quantitative ELISA kit for the measurement of Phosphotyrosine Tec in Human in cell Lysate samples.