Human Calprotectin (S100A8/S100A9) ELISA Kit
Supplier: Abcam
New Product
Certificates
About this item
Human Calprotectin (S100A8/S100A9) ELISA kit is a single-wash 90-min Simplestep used to quantify Human Calprotectin (S100A8/S100A9) with a sensitivity of 13,624 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders - Design your own immunoassay: we also offer the conjugation-ready antibody pair.
Frequently Bought Together