Rat CXCL1 ELISA Kit (GRO alpha)
Catalog # ABCAAB219044-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat CXCL1 ELISA kit (GRO alpha)
Specifications
About this item
Rat CXCL1 ELISA kit (GRO alpha) is a Sandwich (quantitative) ELISA kit for the measurement of Rat CXCL1 (GRO alpha) in Rat in cell culture extracts, Heparin plasma, Citrate plasma, cell culture supernatant, serum, EDTA plasma samples.