Order Entry
Czech Republic
ContactUsLinkComponent
Human Granzyme B ELISPOT Kit (with un-coated plates)
Human Granzyme B ELISPOT Kit (with un-coated plates)
Catalog # ABCAAB46617-480
Supplier:  Abcam
CAS Number:  
undefined
Human Granzyme B ELISPOT Kit (with un-coated plates)
Catalog # ABCAAB46617-480
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Human Granzyme B ELISPOT kit (with un-coated plates)
  • Seq:
    1
  • Species reactivity:
    Human
  • Sample type:
    Suspension cells
  • Form:
    Uncoated
  • Storage temperature:
    +4 °C
  • Processing time:
    Multiple steps
  • Antigen symbol:
    Granzyme B
  • Regulatory status:
    RUO

Specifications

About this item

Human Granzyme B ELISPOT Kit (with un-coated plates) is a ELISPOT kit for the sensitive detection of Human Granzyme B production in a single cell suspension.