Mouse alpha Fetoprotein ELISA Kit
ABCAAB210969-96
: Abcam
:
- Description:Mouse alpha Fetoprotein ELISA kit
Mouse alpha Fetoprotein ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse alpha Fetoprotein with a sensitivity of 287 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.