Rat Haptoglobin ELISA Kit
Catalog # ABCAAB108857-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat Haptoglobin ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Rat Haptoglobin ELISA Kit is a Competitive ELISA kit for the measurement of Rat Haptoglobin in Rat in Plasma, Serum samples.