ContactUsLinkComponent
Human Anti-Herpes simplex virus Type 1 IgG ELISA Kit (HSV1)
  ABCAAB108737-96
 :  Abcam
 :  
undefined
Human Anti-Herpes simplex virus Type 1 IgG ELISA Kit (HSV1)
  ABCAAB108737-96
 :  Abcam
 :  
  • Description:
    Human Anti-Herpes simplex virus Type 1 IgG ELISA Kit (HSV1)
  • Tests per kit:
    96 tests
Human Anti-Herpes simplex virus Type 1 IgG ELISA Kit (HSV1) is a Indirect ELISA kit for the measurement of Human Anti-Herpes simplex virus Type 1 IgG (HSV1) in Human in Citrate plasma, Heparin Plasma, Serum samples.