Canine GM-CSF ELISA Kit
Catalog # ABCAAB273187-1X96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:GM-CSF
- Description:Canine GM-CSF ELISA kit
- Sample type:Plasma, cell culture supernatant, serum
- Detection method:Colorimetric
- Detection range:0,102 - 25 ng/ml
- Storage temperature:4 °C
- Sensitivity:0,13 ng/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Canine GM-CSF ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Canine GM-CSF in Dog in plasma, cell culture supernatant, Serum samples.