ContactUsLinkComponent
Mouse IL-27 p28 ELISA Kit
Mouse IL-27 p28 ELISA Kit
Catalog # ABCAAB171333-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse IL-27 p28 ELISA Kit
Catalog # ABCAAB171333-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Mouse IL-27 p28 ELISA kit
  • Tests per kit:
    96 tests
Specifications
About this item
Mouse IL-27 p28 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse IL-27 p28 in Mouse in Plasma, Cell culture media, Serum samples.