Order Entry
Czech Republic
ContactUsLinkComponent
Mouse IL-6 ELISA Kits
Mouse IL-6 ELISA Kits
Supplier:  Abcam
New Product
Certificates
undefined
Mouse IL-6 ELISA Kits
Supplier:  Abcam
New Product

Specifications

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Target protein:
    IL-6
  • Sample type:
    Cell culture supernatant, citrate plasma, edta plasma, heparin plasma, serum
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Detection range:
    15,6 - 1000 pg/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    11,3 pg/ml
  • Regulatory status:
    RUO

Specifications

Frequently Bought Together