Rat IFN gamma ELISA Kit
Catalog # ABCAAB113349-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat IFN gamma ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Rat IFN gamma ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat IFN gamma in Rat in Plasma, Cell culture supernatant samples.