Human Methotrexate ELISA Kit
Catalog # ABCAAB213976-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Methotrexate ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Human Methotrexate ELISA kit ab213976 is used to quantify Human Methotrexate with a sensitivity of 0,087 ng/ml. Indirect ELISA kit.