Mouse TIMP1 ELISA Kit
ABCAAB100745-96
: Abcam
:
- Description:Mouse TIMP1 ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Mouse TIMP1 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse TIMP1 in Mouse in cell culture extracts, tissue extracts samples.