Mouse c-Kit ELISA Kit (CD117)
ABCAAB203361-96
: Abcam
:
- Description:Mouse c-kit ELISA kit (CD117)
Mouse c-kit ELISA kit (CD117) is a single-wash 90-min Simplestep used to quantify Mouse c-kit (CD117) with a sensitivity of 30 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.