Human Hsp90 alpha ELISA Kit
Supplier: Abcam
New Product
Certificates
About this item
Human Hsp90 alpha ELISA kit is a single-wash 90-min Simplestep used to quantify Human Hsp90 alpha with a sensitivity of 0,29 ng/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 10×96 plates for bulk orders.
Frequently Bought Together