Substance P ELISA Kit
Catalog # ABCAAB288318-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Substance P ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Substance P ELISA Kit ab288318 is used to quantify Substance P with a sensitivity of 0,8445 ng/ml. Competitive ELISA Kit.