Mouse OX40 ELISA Kit (TNFRSF4)
Catalog # ABCAAB213894-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse OX40 ELISA kit (TNFRSF4)
- Tests per kit:96 tests
Specifications
About this item
Mouse OX40 ELISA Kit (TNFRSF4) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse OX40 (TNFRSF4) in Mouse in Cell Lysate, Cell culture supernatant, EDTA Plasma, Heparin Plasma, Serum, Tissue Homogenate samples.