Mouse L-Selectin ELISA Kit (CD62L)
ABCAAB155448-96
: Abcam
:
- Description:Mouse L-Selectin ELISA kit (CD62L)
- Tests per kit:96 tests
Mouse L-Selectin ELISA kit (CD62L) is a sandwich (quantitative) ELISA kit for the measurement of Mouse L-Selectin (CD62L) in mouse in plasma, cell culture supernatant, serum samples.