GRB2 Antibody Pair - BSA and Azide Free
Catalog # ABCAAB315400-10X96
Supplier: Abcam
New Product
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:GRB2
- Description:GRB2 antibody pair - BSA and azide free
- Detection range:0,31 - 20 ng/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:10×96 tests
Specifications
About this item
GRB2 antibody pair - BSA and Azide free.