Order Entry
Czech Republic
ContactUsLinkComponent
Human IL-10 Antibody Pair - BSA and Azide Free (Interleukin-10)
Human IL-10 Antibody Pair - BSA and Azide Free (Interleukin-10)
Catalog # ABCAAB243976-10X96
Supplier:  Abcam
CAS Number:  
undefined
Human IL-10 Antibody Pair - BSA and Azide Free (Interleukin-10)
Catalog # ABCAAB243976-10X96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Format:
    Uncoated
  • Target protein:
    IL-10
  • Description:
    Human IL-10 antibody pair - BSA and azide free (Interleukin-10)
  • Detection range:
    6,86 - 5000 pg/ml
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Tests per kit:
    10×96 tests

Specifications

About this item

Human IL-10 antibody pair - BSA and Azide free (Interleukin-10) is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human IL-10 (Interleukin-10).