Order Entry
Czech Republic
ContactUsLinkComponent
 
Human Pro-Collagen I C-Terminal Propeptide Antibody Pair - BSA and Azide Free (PICP)
Catalog # ABCAAB253334-10X96
Supplier:  Abcam
CAS Number:  
undefined
Human Pro-Collagen I C-Terminal Propeptide Antibody Pair - BSA and Azide Free (PICP)
Catalog # ABCAAB253334-10X96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Format:
    Uncoated
  • Target protein:
    PICP
  • Description:
    Human Pro-Collagen I C-Terminal Propeptide antibody pair - BSA and azide free (PICP)
  • Detection range:
    23,4375 - 1500 pg/ml
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Tests per kit:
    10×96 tests

Specifications

About this item

Human Pro-Collagen I C-Terminal Propeptide antibody pair - BSA and Azide free (PICP) is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Pro-Collagen I C-Terminal Propeptide (PICP).