Mouse DPPIV ELISA Kit (CD26)
Catalog # ABCAAB155455-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse DPPIV ELISA kit (CD26)
- Tests per kit:96 tests
Specifications
About this item
Mouse DPPIV ELISA kit (CD26) is a sandwich (quantitative) ELISA kit for the measurement of Mouse DPPIV (CD26) in mouse in plasma, cell culture supernatant, serum samples.