ContactUsLinkComponent
Human Anti-Respiratory syncytial virus IgM ELISA Kit (RSV)
Human Anti-Respiratory syncytial virus IgM ELISA Kit (RSV)
Catalog # ABCAAB108766-96
Supplier:  Abcam
CAS Number:  
undefined
Human Anti-Respiratory syncytial virus IgM ELISA Kit (RSV)
Catalog # ABCAAB108766-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Human Anti-Respiratory syncytial virus IgM ELISA kit (RSV)
  • Tests per kit:
    96 tests
Specifications
About this item
Human Anti-Respiratory syncytial virus IgM ELISA kit (RSV) is a Indirect ELISA kit for the measurement of Human Anti-Respiratory syncytial virus IgM (RSV) in Human in Citrate plasma, Heparin Plasma, serum samples.