ContactUsLinkComponent
STAT3 pY705 ELISA Kit
STAT3 pY705 ELISA Kit
Catalog # ABCAAB176654-96
Supplier:  Abcam
CAS Number:  
undefined
STAT3 pY705 ELISA Kit
Catalog # ABCAAB176654-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    STAT3 pY705 ELISA kit
  • Tests per kit:
    96 tests
Specifications
About this item
STAT3 pY705 ELISA Kit is a single-wash 90-min Simplestep used to quantify STAT3 pY705 with a sensitivity of 30 µg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.