Rat IL-17F ELISA Kit
Catalog # ABCAAB213913-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat IL-17F ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Rat IL-17F ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Rat IL-17F in Rat in Cell Lysate, cell culture supernatant, Citrate plasma, EDTA Plasma, Heparin Plasma, Serum, Tissue Homogenate samples.