ContactUsLinkComponent
Rat alpha 2 Macroglobulin ELISA Kit
Rat alpha 2 Macroglobulin ELISA Kit
Catalog # ABCAAB157730-96
Supplier:  Abcam
CAS Number:  
undefined
Rat alpha 2 Macroglobulin ELISA Kit
Catalog # ABCAAB157730-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Rat alpha 2 Macroglobulin ELISA kit
Specifications
About this item
Rat alpha 2 Macroglobulin ELISA kit ab157730 is used to quantify Rat alpha 2 Macroglobulin with a sensitivity of 1.795 ng/ml. Sandwich ELISA kit.