ContactUsLinkComponent
Mouse IL36 alpha/IL-1F6 Antibody Pair - BSA and Azide free
Mouse IL36 alpha/IL-1F6 Antibody Pair - BSA and Azide free
Catalog # ABCAAB253820-10X96
Supplier:  Abcam
CAS Number:  
undefined
Mouse IL36 alpha/IL-1F6 Antibody Pair - BSA and Azide free
Catalog # ABCAAB253820-10X96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Mouse IL36 alpha/IL-1F6 antibody pair - BSA and Azide free
  • Tests per kit:
    10×96 tests
Specifications
About this item
Mouse IL36 alpha/IL-1F6 Antibody pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Mouse IL36 alpha/IL-1F6.