Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Rabbit IgM ELISA Kit

Rabbit IgM ELISA Kit

Rabbit IgM ELISA Kit, 96 tests
Catalog # ABCAAB190539-96
Supplier:   Abcam
Rabbit IgM ELISA Kit
Catalog # ABCAAB190539-96
Supplier:  Abcam
CAS Number:  
Rabbit IgM ELISA Kit
Rabbit IgM ELISA Kit

Rabbit IgM ELISA Kit, 96 tests

CAS Number:  
Price
20 220,00 Kč
Log in for availability
1 * 96 Tests
Product Details & Documents

Rabbit IgM ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Rabbit IgM in Rabbit in plasma, serum samples.

Specifications

Specifications

  • Description:
    Rabbit IgM ELISA Kit
  • Tests per kit:
    96 tests
  • Cat. No.:
    ABCAAB190539-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    1,685 ng/ml
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rabbit
  • Sample type:
    Plasma, Serum
  • Target protein:
    IgM
  • Detection range:
    6,25 - 200 ng/ml
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Assay Type:
    Sandwich

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.