Order Entry
Czech Republic
ContactUsLinkComponent
Mouse FGF-21 ELISA Kit
Mouse FGF-21 ELISA Kit
Catalog # ABCAAB212160-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse FGF-21 ELISA Kit
Catalog # ABCAAB212160-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Target protein:
    FGF21
  • Description:
    Mouse FGF-21 ELISA kit
  • Sample type:
    Heparin plasma, citrate plasma, cell culture supernatant, serum, edta plasma
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Detection range:
    9,375 - 600 pg/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    1,47 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Mouse FGF-21 ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse FGF-21 with a sensitivity of 1.47 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Cited in over 20 citations.