Mouse TNF alpha ELISA Kit
Catalog # ABCAAB108910-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse TNF alpha ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Mouse TNF alpha ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse TNF alpha in Mouse in plasma, serum, cell culture supernatant, tissue, cell lysate samples.