Human HSD17B14 ELISA Kit
Catalog # ABCAAB313972-96TST
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:HSD17B14
- Description:Human HSD17B14 ELISA kit
- Detection method:Colorimetric
- Detection range:22 - 5000 pg/ml
- Storage temperature:4 °C
- Sensitivity:22 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Human HSD17B14 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human HSD17B14 in Human in Serum, plasma, cell culture supernatant samples.