ContactUsLinkComponent
Mouse IP-10 Matched Antibody Pair Kit
Mouse IP-10 Matched Antibody Pair Kit
Catalog # ABCAAB275364-5X96
Supplier:  Abcam
CAS Number:  
undefined
Mouse IP-10 Matched Antibody Pair Kit
Catalog # ABCAAB275364-5X96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Mouse IP-10 Matched Antibody Pair Kit
  • Tests per kit:
    5×96 tests
Specifications
About this item
Mouse IP-10 Matched Antibody Pair Kit is a kit for ELISA development for the measurement of Mouse IP-10 by sandwich ELISA method.