Human H2A.X (phospho S139) In-Cell ELISA Kit (IR)
Catalog # ABCAAB131382-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human H2A.X (phospho S139) In-Cell ELISA Kit (IR)
- Tests per kit:96 tests
Specifications
About this item
Human H2A.X (phospho S139) In-Cell ELISA Kit (IR) is Cell-based (qualitative) ELISA for the measurement of Human H2A.X (phospho S139) (IR) production in Human in Suspension cells, Adherent cells samples.