Rabbit CXCL13 ELISA Kit (BCA-1)
ABCAAB273248-1X96
: Abcam
:
- Description:Rabbit CXCL13 ELISA kit (BCA-1)
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Rabbit CXCL13 ELISA kit (BCA-1) is a Sandwich (quantitative) ELISA kit for the measurement of Rabbit CXCL13 (BCA-1) in Rabbit in plasma, cell culture supernatant, serum samples.