Order Entry
Czech Republic
ContactUsLinkComponent
Rat IL-13 Antibody Pair - BSA and Azide free
Rat IL-13 Antibody Pair - BSA and Azide free
Catalog # ABCAAB270384-10X96
Supplier:  Abcam
CAS Number:  
undefined
Rat IL-13 Antibody Pair - BSA and Azide free
Catalog # ABCAAB270384-10X96
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Rat IL-13 antibody pair - BSA and Azide free
  • Storage temperature:
    +4 ℃
  • Tests per kit:
    10×96 tests

Specifications

About this item

Rat IL-13 Antibody Pair - BSA and Azide free.